COMH4211B7B2 Combi-Blocks, Inc.

2-Fluoro-5-(2-methoxyethoxy)pyridine

  • MDL number MFCD24190787
  • Cas Number : 1209778-75-5
  • Molecular Weight : 171.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4811

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4811
Availability details for
Quantity
  • Availability
  • Add to Cart