COMH4211B7B3 Combi-Blocks, Inc.

2-(4-Methoxy-benzylamino)-isonicotinic acid methyl ester

  • MDL number MFCD31557859
  • Cas Number : 1917789-59-3
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4819

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4819
Availability details for
Quantity
  • Availability
  • Add to Cart