COMH4211B7B5 Combi-Blocks, Inc.

2-Methoxyethyl (3-ethynylphenyl)carbamate

  • MDL number MFCD31579400
  • Cas Number : 1816945-63-7
  • Molecular Weight : 219.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4829

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4829
Availability details for
Quantity
  • Availability
  • Add to Cart