COMH4211B7B8 Combi-Blocks, Inc.

(4-Bromo-2-methyl-phenyl)-pyrrolidin-1-yl-methanone

  • MDL number MFCD20543576
  • Cas Number : 276678-35-4
  • Molecular Weight : 268.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4845

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4845
Availability details for
Quantity
  • Availability
  • Add to Cart