COMH4211B7BB Combi-Blocks, Inc.

2-Methyl-4'-(trifluoromethyl)biphenyl-4-carbonitrile

  • MDL number MFCD28579887
  • Cas Number : 1383798-19-3
  • Molecular Weight : 261.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4850

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4850
Availability details for
Quantity
  • Availability
  • Add to Cart