COMH4211B7BC Combi-Blocks, Inc.

[3-Chloro-2-(propan-2-yloxy)phenyl]methanamine

  • MDL number MFCD26055697
  • Cas Number : 918812-03-0
  • Molecular Weight : 199.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4853

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4853
Availability details for
Quantity
  • Availability
  • Add to Cart