COMH4211B7BF Combi-Blocks, Inc.

4-Bromo-3-(hydroxymethyl)-2-methylphenol

  • MDL number MFCD24729792
  • Cas Number : 1255206-72-4
  • Molecular Weight : 217.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4861

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4861
Availability details for
Quantity
  • Availability
  • Add to Cart