COMH4211B7C0 Combi-Blocks, Inc.

3-Fluoro-4-(morpholine-4-carbonyl)aniline

  • MDL number MFCD16084598
  • Cas Number : 934601-04-4
  • Molecular Weight : 224.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4862

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4862
Availability details for
Quantity
  • Availability
  • Add to Cart