COMH4211B7C1 Combi-Blocks, Inc.

2-Bromo-3-(2-methoxy-ethoxy)-benzylamine

  • MDL number MFCD31613855
  • Cas Number : 2025971-96-2
  • Molecular Weight : 260.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4863

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4863
Availability details for
Quantity
  • Availability
  • Add to Cart