COMH4211B7C4 Combi-Blocks, Inc.

3-Chloro-4-cyclopropylaniline

  • MDL number MFCD14585518
  • Cas Number : 1208082-73-8
  • Molecular Weight : 167.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4877

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4877
Availability details for
Quantity
  • Availability
  • Add to Cart