COMH4211B7C5 Combi-Blocks, Inc.

4-Methyl-5-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyridine-2-carboxylic acid diethylamide

  • MDL number MFCD31613904
  • Molecular Weight : 318.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4878

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4878
Availability details for
Quantity
  • Availability
  • Add to Cart