COMH4211B7C6 Combi-Blocks, Inc.

5-Chloro-pyrazine-2-carboxylic acid (2,2,2-trifluoro-ethyl)-amide

  • MDL number MFCD31381728
  • Cas Number : 1691930-69-4
  • Molecular Weight : 239.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4879

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4879
Availability details for
Quantity
  • Availability
  • Add to Cart