COMH4211B7C9 Combi-Blocks, Inc.

(3-Bromo-4-(2,4-difluorophenoxy)phenyl)methanol

  • MDL number MFCD31614037
  • Cas Number : 1446236-25-4
  • Molecular Weight : 315.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4883

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4883
Availability details for
Quantity
  • Availability
  • Add to Cart