COMH4211B7CA Combi-Blocks, Inc.

2-(4-Trifluoromethyl-phenoxy)-propan-1-ol

  • MDL number MFCD31579458
  • Cas Number : 69183-54-6
  • Molecular Weight : 220.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4-(trifluoromethyl)phenoxy)propan-1-ol
Inchi Code 1S/C10H11F3O2/c1-7(6-14)15-9-4-2-8(3-5-9)10(11,12)13/h2-5,7,14H,6H2,1H3
Inchi Key RXMPMXVVRWRGMZ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4888

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4888
Availability details for
Quantity
  • Availability
  • Add to Cart