COMH4211B7CC Combi-Blocks, Inc.

3-Chloro-n-(1,1-dimethylethyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-benzamide

  • MDL number MFCD28718045
  • Cas Number : 1442432-30-5
  • Molecular Weight : 337.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4890

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4890
Availability details for
Quantity
  • Availability
  • Add to Cart