COMH4211B7CD Combi-Blocks, Inc.

2-(Cyclopentyloxy)phenol

  • MDL number MFCD11103434
  • Cas Number : 29941-90-0
  • Molecular Weight : 178.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4891

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4891
Availability details for
Quantity
  • Availability
  • Add to Cart