COMH4211B7CE Combi-Blocks, Inc.

(12-Bromo-dodecyloxy)-tert-butyl-dimethyl-silane

  • MDL number MFCD31561453
  • Cas Number : 134413-60-8
  • Molecular Weight : 379.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ((12-bromododecyl)oxy)(tert-butyl)dimethylsilane
Inchi Code 1S/C18H39BrOSi/c1-18(2,3)21(4,5)20-17-15-13-11-9-7-6-8-10-12-14-16-19/h6-17H2,1-5H3
Inchi Key NFUQZVUAJDEGGF-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4895

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4895
Availability details for
Quantity
  • Availability
  • Add to Cart