COMH4211B7D2 Combi-Blocks, Inc.

4-Amino-n,n-diethyl-2-methyl-benzamide

  • MDL number MFCD16117677
  • Cas Number : 1188506-56-0
  • Molecular Weight : 206.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4900

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4900
Availability details for
Quantity
  • Availability
  • Add to Cart