COMH4211B7D4 Combi-Blocks, Inc.

4-(4-Chloro-3-trifluoromethyl-phenoxy)-benzoic acid

  • MDL number MFCD31579438
  • Cas Number : 1354960-58-9
  • Molecular Weight : 316.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4903

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4903
Availability details for
Quantity
  • Availability
  • Add to Cart