COMH4211B7D5 Combi-Blocks, Inc.

(2-Chloro-4-cyclopropylphenyl)methanol

  • MDL number MFCD31583193
  • Cas Number : 1610416-51-7
  • Molecular Weight : 182.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4904

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4904
Availability details for
Quantity
  • Availability
  • Add to Cart