COMH4211B7D6 Combi-Blocks, Inc.

Ethyl 2-(4-aminophenyl)-2-methylpropanoate

  • MDL number MFCD23378786
  • Cas Number : 36924-81-9
  • Molecular Weight : 207.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4907

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4907
Availability details for
Quantity
  • Availability
  • Add to Cart