COMH4211B7D8 Combi-Blocks, Inc.

R Piperidin-3-yl-pyrrolidin-1-yl-methanone, hydrochloride

  • MDL number MFCD30830198
  • Cas Number : 1469389-13-6
  • Molecular Weight : 218.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4912

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4912
Availability details for
Quantity
  • Availability
  • Add to Cart