COMH4211B7DA Combi-Blocks, Inc.

5-[3-(Trifluoromethyl)phenyl]thiophene-2-carboxylic acid

  • MDL number MFCD06801924
  • Cas Number : 893733-18-1
  • Molecular Weight : 272.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4919

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4919
Availability details for
Quantity
  • Availability
  • Add to Cart