COMH4211B7DB Combi-Blocks, Inc.

Trans (4-hydroxy-cyclohexyl)-carbamic acid allyl ester

  • MDL number MFCD22581341
  • Cas Number : 1391573-83-3
  • Molecular Weight : 199.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4920

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4920
Availability details for
Quantity
  • Availability
  • Add to Cart