COMH4211B7DC Combi-Blocks, Inc.

tert-Butyl (4-bromobutyloxy)acetate

  • MDL number MFCD25963657
  • Cas Number : 475086-55-6
  • Molecular Weight : 267.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4925

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4925
Availability details for
Quantity
  • Availability
  • Add to Cart