COMH4211B7DD Combi-Blocks, Inc.

1-(O-Tolyloxy)-2-iodobenzene

  • MDL number MFCD31583023
  • Cas Number : 884512-16-7
  • Molecular Weight : 310.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4927

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4927
Availability details for
Quantity
  • Availability
  • Add to Cart