COMH4211B7E0 Combi-Blocks, Inc.

1-Cyclobutyl-6-cyclopropyl-1h-indole-3-carbonitrile

  • MDL number MFCD31583007
  • Cas Number : 1248585-28-5
  • Molecular Weight : 236.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4938

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4938
Availability details for
Quantity
  • Availability
  • Add to Cart