COMH4211B7E2 Combi-Blocks, Inc.

2-Methoxy-4-(2-methyl-imidazol-1-yl)-phenylamine

  • MDL number MFCD09029883
  • Cas Number : 218301-72-5
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4941

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4941
Availability details for
Quantity
  • Availability
  • Add to Cart