COMH4211B7E4 Combi-Blocks, Inc.

3-((1R,4R)-4-Aminocyclohexyl)propanenitrile

  • MDL number MFCD30530593
  • Cas Number : 1315498-83-9
  • Molecular Weight : 152.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4946

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4946
Availability details for
Quantity
  • Availability
  • Add to Cart