COMH4211B7E5 Combi-Blocks, Inc.

(R)-2-Amino-4-methyl-1-(pyrrolidin-1-yl)pentan-1-one

  • MDL number MFCD20545771
  • Cas Number : 1258066-52-2
  • Molecular Weight : 184.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4947

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4947
Availability details for
Quantity
  • Availability
  • Add to Cart