COMH4211B7E6 Combi-Blocks, Inc.

N-Succinimidyl-s-acetylthiohexanoate

  • MDL number MFCD23103970
  • Cas Number : 874743-76-7
  • Molecular Weight : 287.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4948

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4948
Availability details for
Quantity
  • Availability
  • Add to Cart