COMH4211B7E8 Combi-Blocks, Inc.

2-(tert-Butylamino)-4'-chloropropiophenone hydrochloride

  • MDL number MFCD01696537
  • Cas Number : 1049718-72-0
  • Molecular Weight : 276.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4950

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4950
Availability details for
Quantity
  • Availability
  • Add to Cart