COMH4211B7E9 Combi-Blocks, Inc.

2-Methyl-3-(2-methylpropoxy)benzoic acid

  • MDL number MFCD24230905
  • Cas Number : 339544-62-6
  • Molecular Weight : 208.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4953

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4953
Availability details for
Quantity
  • Availability
  • Add to Cart