COMH4211B7EA Combi-Blocks, Inc.

3-Bromo-2-fluoro-n-methylaniline

  • MDL number MFCD09745407
  • Cas Number : 943830-86-2
  • Molecular Weight : 204.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4954

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4954
Availability details for
Quantity
  • Availability
  • Add to Cart