COMH4211B7EC Combi-Blocks, Inc.

1-[3-(2,2-Difluoro-ethoxy)-phenyl]-ethylamine hydrochloride

  • MDL number MFCD31381725
  • Cas Number : 2205504-95-4
  • Molecular Weight : 237.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4956

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4956
Availability details for
Quantity
  • Availability
  • Add to Cart