COMH4211B7ED Combi-Blocks, Inc.

(4-Bromo-thiophen-2-ylmethyl)-cyclopropyl-carbamic acid tert-butyl ester

  • MDL number MFCD31613870
  • Cas Number : 2270905-79-6
  • Molecular Weight : 332.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4958

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4958
Availability details for
Quantity
  • Availability
  • Add to Cart