COMH4211B7EE Combi-Blocks, Inc.

1-(1-Phenyl-ethyl)-1h-pyrazol-4-ol

  • MDL number MFCD31561480
  • Cas Number : 1629171-80-7
  • Molecular Weight : 188.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4959

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4959
Availability details for
Quantity
  • Availability
  • Add to Cart