COMH4211B7F1 Combi-Blocks, Inc.

4-(1-tert-Butoxycarbonylpyrrolidin-3-yloxy)benzaldehyde

  • MDL number MFCD24469837
  • Cas Number : 247132-69-0
  • Molecular Weight : 291.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4966

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4966
Availability details for
Quantity
  • Availability
  • Add to Cart