COMH4211B7F3 Combi-Blocks, Inc.

4-Benzyloxy-3,3-dimethyl-butyric acid

  • MDL number MFCD29924179
  • Molecular Weight : 222.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4972

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4972
Availability details for
Quantity
  • Availability
  • Add to Cart