COMH4211B7F4 Combi-Blocks, Inc.

(2S)-2-Amino-n,4-dimethylpentanamide

  • MDL number MFCD14696986
  • Cas Number : 64569-68-2
  • Molecular Weight : 144.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4973

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4973
Availability details for
Quantity
  • Availability
  • Add to Cart