COMH4211B7F5 Combi-Blocks, Inc.

1-(4-(2-Bromoethoxy)phenyl)ethanone

  • MDL number MFCD01126777
  • Cas Number : 55368-24-6
  • Molecular Weight : 243.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4976

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4976
Availability details for
Quantity
  • Availability
  • Add to Cart