COMH4211B7F8 Combi-Blocks, Inc.

N-(2-Methylbenzyl)propane-1,3-diamine

  • MDL number MFCD12765761
  • Cas Number : 38486-29-2
  • Molecular Weight : 178.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4992

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4992
Availability details for
Quantity
  • Availability
  • Add to Cart