COMH4211B7FA Combi-Blocks, Inc.

6-Bromo-2-phenyl-2h-pyrazolo[4,3-b]pyridin-3-amine

  • MDL number MFCD31561609
  • Cas Number : 1811512-33-0
  • Molecular Weight : 289.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4994

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4994
Availability details for
Quantity
  • Availability
  • Add to Cart