COMH4211B7FC Combi-Blocks, Inc.

5-Bromo-2-(4-fluoro-phenylsulfanyl)-nicotinic acid tert-butyl ester

  • MDL number MFCD31557893
  • Cas Number : 2206609-39-2
  • Molecular Weight : 384.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4996

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4996
Availability details for
Quantity
  • Availability
  • Add to Cart