COMH4211B7FD Combi-Blocks, Inc.

2,6-Naphthalenediacetic acid

  • MDL number MFCD09925192
  • Cas Number : 128822-58-2
  • Molecular Weight : 244.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4997

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4997
Availability details for
Quantity
  • Availability
  • Add to Cart