COMH4211B7FE Combi-Blocks, Inc.

(4-(Pyridin-2-yloxymethyl)-phenyl)-methanol

  • MDL number MFCD31579358
  • Cas Number : 936342-24-4
  • Molecular Weight : 215.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4998

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4998
Availability details for
Quantity
  • Availability
  • Add to Cart