COMH4211B801 Combi-Blocks, Inc.

3-[2-(4-Fluorophenyl)ethoxy]propanoic acid

  • MDL number MFCD19370495
  • Cas Number : 189012-54-2
  • Molecular Weight : 212.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5009

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5009
Availability details for
Quantity
  • Availability
  • Add to Cart