COMH4211B802 Combi-Blocks, Inc.

N-(2-Aminoethyl)-5-methylthiophene-2-carboxamide

  • MDL number MFCD09805994
  • Cas Number : 103879-56-7
  • Molecular Weight : 184.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5013

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5013
Availability details for
Quantity
  • Availability
  • Add to Cart