COMH4211B803 Combi-Blocks, Inc.

tert-Butyl 4-(4-amino-2-chlorophenoxy)piperidine-1-carboxylate

  • MDL number MFCD27979532
  • Cas Number : 337519-87-6
  • Molecular Weight : 326.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5014

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5014
Availability details for
Quantity
  • Availability
  • Add to Cart