COMH4211B805 Combi-Blocks, Inc.

(1-Methanesulfonylpiperidin-2-yl)methanol

  • MDL number MFCD01212955
  • Cas Number : 349403-28-7
  • Molecular Weight : 193.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [1-(methylsulfonyl)-2-piperidinyl]methanol
Inchi Code 1S/C7H15NO3S/c1-12(10,11)8-5-3-2-4-7(8)6-9/h7,9H,2-6H2,1H3
Inchi Key FLCRKCCKSJQLQA-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5017

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5017
Availability details for
Quantity
  • Availability
  • Add to Cart